Product Details

SNP ID
rs150667938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30319330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTGCTACTAGCAAAGAAGCAA[A/C]AGTCCATGGAGTTTGAATTTTTGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP26-1 PubMed Links

Gene Details

Gene
KRTAP26-1
Gene Name
keratin associated protein 26-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203405.1 960 UTR 3 NP_981950.1

View Full Product Details