Product Details

SNP ID
rs138582408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50253441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGAGACCCTGGCCCCCAGCTGC[C/T]GGGGAGGCTTGAAGCTGAGCACCTC
Phenotype
MIM: 608544 MIM: 602399 MIM: 610053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDAC10 PubMed Links

Gene Details

Gene
HDAC10
Gene Name
histone deacetylase 10
There are no transcripts associated with this gene.

Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303252.1 1284 Missense Mutation CAG,CGG Q345R NP_001290181.1
NM_002969.4 1284 Missense Mutation CAG,CGG Q355R NP_002960.2
Gene
TUBGCP6
Gene Name
tubulin gamma complex associated protein 6
There are no transcripts associated with this gene.

View Full Product Details