Product Details

SNP ID
rs138944919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50569712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGAAGGCATAAATCAAACCTT[A/G]AAGATCTGAAAGCCAAGCTGATATT
Phenotype
MIM: 612395 MIM: 601987 MIM: 615775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKB PubMed Links

Gene Details

Gene
CHKB
Gene Name
choline kinase beta
There are no transcripts associated with this gene.

Gene
CHKB-CPT1B
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CPT1B
Gene Name
carnitine palmitoyltransferase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145134.1 Intron NP_001138606.1
NM_001145135.1 Intron NP_001138607.1
NM_001145137.1 Intron NP_001138609.1
NM_004377.3 Intron NP_004368.1
NM_152245.2 Intron NP_689451.1
NM_152246.2 Intron NP_689452.1
Gene
SYCE3
Gene Name
synaptonemal complex central element protein 3
There are no transcripts associated with this gene.

View Full Product Details