Product Details

SNP ID
rs139840200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39015670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTATGGGAAGCCAACGATCGGAA[C/T]GAAACTTGGCTGTGCTTCACCGTGG
Phenotype
MIM: 607750 MIM: 609900 MIM: 608993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOBEC3C PubMed Links

Gene Details

Gene
APOBEC3C
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014508.2 196 Silent Mutation AAC,AAT N31N NP_055323.2
Gene
APOBEC3D
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3D
There are no transcripts associated with this gene.

Gene
APOBEC3F
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
There are no transcripts associated with this gene.

View Full Product Details