Product Details
- SNP ID
-
rs141562718
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:16964821 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGTCTGTGTTGGTGATGCTCTGC[A/G]TGGAGATGGGGTGTTGGGAGGAGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAB4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5992599] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAB4
- Gene Name
- GRB2 associated binding protein family member 4
View Full Product Details