Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022719.2 | 4057 | UTR 3 | NP_073210.1 | ||
XM_005261282.4 | 4057 | Intron | XP_005261339.1 | ||
XM_006724329.3 | 4057 | Intron | XP_006724392.1 | ||
XM_006724330.3 | 4057 | Intron | XP_006724393.1 | ||
XM_006724331.3 | 4057 | Intron | XP_006724394.1 | ||
XM_011530403.1 | 4057 | Intron | XP_011528705.1 | ||
XM_011530404.2 | 4057 | Intron | XP_011528706.1 | ||
XM_017028958.1 | 4057 | Intron | XP_016884447.1 | ||
XM_017028959.1 | 4057 | Intron | XP_016884448.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_053006.4 | 4057 | Missense Mutation | AGC,GGC | S70G | NP_443732.3 |