Product Details

SNP ID
rs143095407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19149017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCTCCCCTTCGGGGACTTCT[C/T]GACGCCGGGCAGGAGCCTCGCGGAA
Phenotype
MIM: 601755 MIM: 601845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR14 PubMed Links

Gene Details

Gene
DGCR14
Gene Name
DiGeorge syndrome critical region gene 14
There are no transcripts associated with this gene.

Gene
GSC2
Gene Name
goosecoid homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005315.1 592 Missense Mutation AAG,GAG K198E NP_005306.1

View Full Product Details