Product Details

SNP ID
rs143245989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43132918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTTTATGTGGCCTTCAAAGCA[C/T]GTTGCAAACAAATCCCTTTCACTCC
Phenotype
MIM: 603392 MIM: 614479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIK PubMed Links

Gene Details

Gene
BIK
Gene Name
BCL2 interacting killer
There are no transcripts associated with this gene.

Gene
MCAT
Gene Name
malonyl-CoA-acyl carrier protein transacylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014507.3 1347 UTR 3 NP_055322.1
NM_173467.4 1347 UTR 3 NP_775738.3
XM_011530156.2 1347 UTR 3 XP_011528458.1

View Full Product Details