Product Details

SNP ID
rs148167797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41808914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGGACCTTCTTCAATTCCTGGC[A/C]TTCCTCTTTGTCCTGCTTTTGTCTG
Phenotype
MIM: 608797
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC134 PubMed Links

Gene Details

Gene
CCDC134
Gene Name
coiled-coil domain containing 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304797.1 275 Silent Mutation GCA,GCC A8A NP_001291726.1
NM_024821.3 275 Silent Mutation GCA,GCC A8A NP_079097.1
XM_005261748.3 275 Silent Mutation GCA,GCC A8A XP_005261805.1
Gene
MEI1
Gene Name
meiotic double-stranded break formation protein 1
There are no transcripts associated with this gene.

View Full Product Details