Product Details
- SNP ID
-
rs150514257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:26169732 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATCTCGCTGTTCCTCGCTCTGCT[C/T]CTGGGGAGCCCGGCGGCAGCGCTGG
- Phenotype
-
MIM: 607021
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEZ6L
PubMed Links
Gene Details
- Gene
- SEZ6L
- Gene Name
- seizure related 6 homolog like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184773.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
NP_001171702.1 |
NM_001184774.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
NP_001171703.1 |
NM_001184775.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
NP_001171704.1 |
NM_001184776.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
NP_001171705.1 |
NM_001184777.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
NP_001171706.1 |
NM_021115.4 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
NP_066938.2 |
XM_005261439.3 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_005261496.1 |
XM_005261440.3 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_005261497.1 |
XM_006724195.3 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_006724258.1 |
XM_011530037.2 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_011528339.1 |
XM_011530038.2 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_011528340.1 |
XM_011530039.2 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_011528341.1 |
XM_017028699.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_016884188.1 |
XM_017028700.1 |
259 |
Silent Mutation |
CTC,CTT |
L21L |
XP_016884189.1 |
View Full Product Details