Product Details

SNP ID
rs138496251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:210435391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATAAAGTGTACAAAATACCTTC[A/G]AAGAGAGAGAAAGGGGTGTCTGGTG
Phenotype
MIM: 604155
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LANCL1 PubMed Links

Gene Details

Gene
LANCL1
Gene Name
LanC like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136574.1 1194 Silent Mutation TTC,TTT F373F NP_001130046.1
NM_001136575.1 1194 Silent Mutation TTC,TTT F373F NP_001130047.1
NM_006055.2 1194 Silent Mutation TTC,TTT F373F NP_006046.1
XM_005246243.2 1194 Silent Mutation TTC,TTT F385F XP_005246300.1
Gene
LANCL1-AS1
Gene Name
LANCL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details