Product Details

SNP ID
rs138820115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197572047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGTCACTCCATCTTCCTGATC[A/C]AACTGTCCGTCGTCTAATTCCCGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RFTN2 PubMed Links

Gene Details

Gene
RFTN2
Gene Name
raftlin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144629.2 2086 Missense Mutation TTG,TTT L489F NP_653230.2
XM_011510595.2 2086 Missense Mutation TTG,TTT L499F XP_011508897.1
XM_011510596.2 2086 Missense Mutation TTG,TTT L498F XP_011508898.1
XM_011510597.2 2086 Missense Mutation TTG,TTT L488F XP_011508899.1
XM_011510598.2 2086 Missense Mutation TTG,TTT L482F XP_011508900.1
XM_017003332.1 2086 Missense Mutation TTG,TTT L472F XP_016858821.1

View Full Product Details