Product Details

SNP ID
rs138861686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73732224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAGAGTTCTTGTCTTCATTTTT[C/T]CCAGTTTGTAGAGGTGAACTGCTTT
Phenotype
MIM: 608680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPRKB PubMed Links

Gene Details

Gene
TPRKB
Gene Name
TP53RK binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264343.3 674 Intron XP_005264400.1
XM_006712026.3 674 Missense Mutation GAA,GGA E107G XP_006712089.1
XM_006712027.3 674 Missense Mutation GAA,GGA E107G XP_006712090.1
XM_011532877.2 674 Missense Mutation GAA,GGA E107G XP_011531179.1
XM_011532878.2 674 Intron XP_011531180.1
XM_011532879.2 674 Missense Mutation GAA,GGA E35G XP_011531181.1
XM_011532880.2 674 Missense Mutation GAA,GGA E35G XP_011531182.1
XM_017004232.1 674 Missense Mutation GAA,GGA E107G XP_016859721.1
XM_017004233.1 674 Missense Mutation GAA,GGA E107G XP_016859722.1
XM_017004234.1 674 Missense Mutation GAA,GGA E68G XP_016859723.1
XM_017004235.1 674 Missense Mutation GAA,GGA E68G XP_016859724.1

View Full Product Details