Product Details
- SNP ID
-
rs139037539
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88563139 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTGTGAAAATTTCTTTAGAGATA[C/T]AACATGACATTTGAGAAACTGCCAA
- Phenotype
-
MIM: 604032
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF2AK3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11330283] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF2AK3
- Gene Name
- eukaryotic translation initiation factor 2 alpha kinase 3
- Gene
- LOC101928371
- Gene Name
- uncharacterized LOC101928371
There are no transcripts associated with this gene.
View Full Product Details