Product Details
- SNP ID
-
rs139177410
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:73208478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGGAGGACCTACAGGACACTGAGA[A/G]ACAGCAAAAGAGAGTCCGAACTATG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOTO
PubMed Links
Gene Details
- Gene
- NOTO
- Gene Name
- notochord homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134462.1 |
870 |
Missense Mutation |
AAA,AGA |
K154R |
NP_001127934.1 |
- Gene
- SMYD5
- Gene Name
- SMYD family member 5
There are no transcripts associated with this gene.
View Full Product Details