Product Details

SNP ID
rs139733155
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:63842191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTACTAAGCCCCTAAAATGTC[A/G]AGATTTGTACAAGGTAAGAAATGCT
Phenotype
MIM: 191760 MIM: 613580
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC647115 PubMed Links
Additional Information
For this assay, SNP(s) [rs537386784,rs577912232] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC647115
Gene Name
translation initiation factor IF-2
There are no transcripts associated with this gene.

Gene
UGP2
Gene Name
UDP-glucose pyrophosphorylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001521.1 303 Intron NP_001001521.1
NM_006759.3 303 Silent Mutation TCA,TCG S2S NP_006750.3
XM_005264537.2 303 UTR 5 XP_005264594.1
XM_005264538.1 303 Intron XP_005264595.1
XM_006712088.1 303 Intron XP_006712151.1
XM_011533091.1 303 Intron XP_011531393.1
XM_017004855.1 303 Intron XP_016860344.1
XM_017004856.1 303 Intron XP_016860345.1
XM_017004857.1 303 Intron XP_016860346.1
Gene
WDPCP
Gene Name
WD repeat containing planar cell polarity effector
There are no transcripts associated with this gene.

View Full Product Details