Product Details

SNP ID
rs140031618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176093316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACTACAGCTGCCGTATGTCGCA[C/T]GGCGTGGGCTTACAGCAGAATGCGC
Phenotype
MIM: 142991 MIM: 142988 MIM: 142989
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVX2 PubMed Links

Gene Details

Gene
EVX2
Gene Name
even-skipped homeobox 2
There are no transcripts associated with this gene.

Gene
HOXD12
Gene Name
homeobox D12
There are no transcripts associated with this gene.

Gene
HOXD13
Gene Name
homeobox D13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000523.3 513 Silent Mutation CAC,CAT H142H NP_000514.2
XM_011511068.2 513 Intron XP_011509370.1
XM_011511069.2 513 Intron XP_011509371.1

View Full Product Details