Product Details

SNP ID
rs140763514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45006282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCGCCTGGCACCGGCACTGGC[A/G]CTGCGCTGGGGCCCGGAGGGTGGCC
Phenotype
MIM: 604994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIX2 PubMed Links

Gene Details

Gene
SIX2
Gene Name
SIX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016932.4 1057 Missense Mutation GCG,GTG A255V NP_058628.3
XM_005264100.3 1057 Missense Mutation GCG,GTG A257V XP_005264157.1

View Full Product Details