Product Details

SNP ID
rs142594188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162171039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCATGTGGGTGTATAAGTGGTT[C/T]GTGGACAGGCCGGATAAGCCGTGGT
Phenotype
MIM: 600403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAP PubMed Links

Gene Details

Gene
FAP
Gene Name
fibroblast activation protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291807.1 2356 Silent Mutation ACA,ACG T716T NP_001278736.1
NM_004460.3 2356 Silent Mutation ACA,ACG T741T NP_004451.2
XM_011510796.2 2356 Silent Mutation ACA,ACG T731T XP_011509098.1
XM_011510797.2 2356 Intron XP_011509099.1
XM_017003585.1 2356 Intron XP_016859074.1
Gene
LOC101929532
Gene Name
uncharacterized LOC101929532
There are no transcripts associated with this gene.

View Full Product Details