Product Details

SNP ID
rs143482964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99162517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTATTATGTAGTACTGATGGGA[C/T]GTTCTTGTCTTCTTTGCTAAAGAGC
Phenotype
MIM: 610284 MIM: 607166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPT1 PubMed Links

Gene Details

Gene
LIPT1
Gene Name
lipoyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204830.1 658 Missense Mutation ACG,ATG T187M NP_001191759.1
NM_015929.3 658 Missense Mutation ACG,ATG T187M NP_057013.1
NM_145197.2 658 Missense Mutation ACG,ATG T187M NP_660198.1
NM_145198.2 658 Missense Mutation ACG,ATG T187M NP_660199.1
NM_145199.2 658 Missense Mutation ACG,ATG T187M NP_660200.1
Gene
MITD1
Gene Name
microtubule interacting and trafficking domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320417.1 658 Intron NP_001307346.1
NM_001320418.1 658 Intron NP_001307347.1
NM_001320419.1 658 Intron NP_001307348.1
NM_138798.2 658 Intron NP_620153.1
XM_011510581.2 658 Intron XP_011508883.1
XM_011510582.2 658 Intron XP_011508884.1
XM_011510583.1 658 Intron XP_011508885.1
XM_017003314.1 658 Intron XP_016858803.1
XM_017003315.1 658 Intron XP_016858804.1
Gene
TSGA10
Gene Name
testis specific 10
There are no transcripts associated with this gene.

View Full Product Details