Product Details

SNP ID
rs144417465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73730611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGGAAGATTTTTCAGAGAAAC[C/T]TGATGACCTTCTACTTGAGATATTA
Phenotype
MIM: 608680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPRKB PubMed Links

Gene Details

Gene
TPRKB
Gene Name
TP53RK binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264343.3 861 Intron XP_005264400.1
XM_006712026.3 861 Silent Mutation CAA,CAG Q169Q XP_006712089.1
XM_006712027.3 861 Silent Mutation CAA,CAG Q169Q XP_006712090.1
XM_011532877.2 861 Silent Mutation CAA,CAG Q169Q XP_011531179.1
XM_011532878.2 861 Intron XP_011531180.1
XM_011532879.2 861 Silent Mutation CAA,CAG Q97Q XP_011531181.1
XM_011532880.2 861 Silent Mutation CAA,CAG Q97Q XP_011531182.1
XM_017004232.1 861 Silent Mutation CAA,CAG Q169Q XP_016859721.1
XM_017004233.1 861 Silent Mutation CAA,CAG Q169Q XP_016859722.1
XM_017004234.1 861 Silent Mutation CAA,CAG Q130Q XP_016859723.1
XM_017004235.1 861 Silent Mutation CAA,CAG Q130Q XP_016859724.1

View Full Product Details