Product Details

SNP ID
rs145219467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108381827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGTGATGTGGAGAAAAGTAAA[C/T]GGGCACCGACTCATCAACGATTTCC
Phenotype
MIM: 608357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT1C4 PubMed Links

Gene Details

Gene
SULT1C4
Gene Name
sulfotransferase family 1C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321770.1 662 Missense Mutation CGG,TGG R79W NP_001308699.1
NM_006588.3 662 Missense Mutation CGG,TGG R79W NP_006579.2
XM_017003807.1 662 UTR 5 XP_016859296.1

View Full Product Details