Product Details

SNP ID
rs145782994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27277876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAGAAATATTCAAAGAGATCA[A/G]CGCAGCTCATGCCATACTGAGCGAC
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 592 Missense Mutation AAC,AGC N79S NP_001290056.1
NM_001303128.1 592 Intron NP_001290057.1
NM_173650.2 592 Missense Mutation AAC,AGC N79S NP_775921.1
XM_006711995.3 592 Missense Mutation AAC,AGC N79S XP_006712058.1
XM_017003872.1 592 Missense Mutation AAC,AGC N79S XP_016859361.1
XM_017003873.1 592 Missense Mutation AAC,AGC N79S XP_016859362.1
XM_017003874.1 592 Missense Mutation AAC,AGC N79S XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
There are no transcripts associated with this gene.

Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details