Product Details

SNP ID
rs145982613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88557757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTAATTGCTTGGCAAAGGGCT[A/T]TGGGAGTTGTTGGACTGTCTTGAAT
Phenotype
MIM: 604032
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF2AK3 PubMed Links

Gene Details

Gene
EIF2AK3
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313915.1 3632 Silent Mutation CAA,CAT Q959H NP_001300844.1
NM_004836.6 3632 Silent Mutation CAA,CAT Q1110H NP_004827.4
XM_017005376.1 3632 Silent Mutation CAA,CAT Q882H XP_016860865.1
Gene
LOC101928371
Gene Name
uncharacterized LOC101928371
There are no transcripts associated with this gene.

View Full Product Details