Product Details

SNP ID
rs146396158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168871550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCTCAAGATGAGGGGCACTAT[C/T]TGACACTAGAAAAAAAAAAAAAAGA
Phenotype
MIM: 607496 MIM: 609395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOSTRIN PubMed Links

Gene Details

Gene
NOSTRIN
Gene Name
nitric oxide synthase trafficking
There are no transcripts associated with this gene.

Gene
SPC25
Gene Name
SPC25, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020675.3 687 Missense Mutation AAT,GAT N186D NP_065726.1
XM_011511516.2 687 Intron XP_011509818.1

View Full Product Details