Product Details

SNP ID
rs146509505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27124060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTACTACTGGGCATGTGTGCTT[A/C]AGGTGGGTGCGGGTCCACCGCTCTG
Phenotype
MIM: 612195 MIM: 606137 MIM: 606395
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABHD1 PubMed Links

Gene Details

Gene
ABHD1
Gene Name
abhydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032604.3 272 Missense Mutation AAG,CAG K38Q NP_115993.3
XM_011533135.2 272 Missense Mutation AAG,CAG K38Q XP_011531437.1
XM_011533136.2 272 Missense Mutation AAG,CAG K38Q XP_011531438.1
XM_011533137.2 272 Missense Mutation AAG,CAG K38Q XP_011531439.1
XM_011533138.2 272 Missense Mutation TTA,TTC L28F XP_011531440.1
XM_017005113.1 272 Missense Mutation AAG,CAG K38Q XP_016860602.1
Gene
CGREF1
Gene Name
cell growth regulator with EF-hand domain 1
There are no transcripts associated with this gene.

Gene
PREB
Gene Name
prolactin regulatory element binding
There are no transcripts associated with this gene.

View Full Product Details