Product Details

SNP ID
rs148129550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96761433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTCCTCCGGAGGAGCGAGAGCA[C/T]GAAGCTGTATGCACTGTGCACCCGG
Phenotype
MIM: 607805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNNM4 PubMed Links

Gene Details

Gene
CNNM4
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020184.3 449 Missense Mutation ACG,ATG T145M NP_064569.3
XM_005263914.3 449 Missense Mutation ACG,ATG T145M XP_005263971.1
XM_005263915.3 449 Missense Mutation ACG,ATG T145M XP_005263972.1
XM_011510955.2 449 Missense Mutation ACG,ATG T145M XP_011509257.1
XM_011510956.2 449 Missense Mutation ACG,ATG T145M XP_011509258.1
XM_017003799.1 449 Intron XP_016859288.1

View Full Product Details