Product Details

SNP ID
rs148515124
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73730727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGATACCAAATTTTTTCAAAG[C/T]CTCTGAAATCTAAGAGAAAAAAAAT
Phenotype
MIM: 608680
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TPRKB PubMed Links

Gene Details

Gene
TPRKB
Gene Name
TP53RK binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264343.3 745 Intron XP_005264400.1
XM_006712026.3 745 Missense Mutation ACT,GCT T131A XP_006712089.1
XM_006712027.3 745 Missense Mutation ACT,GCT T131A XP_006712090.1
XM_011532877.2 745 Missense Mutation ACT,GCT T131A XP_011531179.1
XM_011532878.2 745 Intron XP_011531180.1
XM_011532879.2 745 Missense Mutation ACT,GCT T59A XP_011531181.1
XM_011532880.2 745 Missense Mutation ACT,GCT T59A XP_011531182.1
XM_017004232.1 745 Missense Mutation ACT,GCT T131A XP_016859721.1
XM_017004233.1 745 Missense Mutation ACT,GCT T131A XP_016859722.1
XM_017004234.1 745 Missense Mutation ACT,GCT T92A XP_016859723.1
XM_017004235.1 745 Missense Mutation ACT,GCT T92A XP_016859724.1

View Full Product Details