Product Details
- SNP ID
-
rs137865812
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108757111 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTATCCATAACGGAGTCTAAGGA[A/G]CACTGAGTACTCCCCGGGTTGATCA
- Phenotype
-
MIM: 605645
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RETNLB
PubMed Links
Gene Details
- Gene
- RETNLB
- Gene Name
- resistin like beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032579.2 |
173 |
Silent Mutation |
TGC,TGT |
C25C |
NP_115968.1 |
View Full Product Details