Product Details

SNP ID
rs137950708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195863561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGGCCTTGGGCCCTGGTCCCC[A/G]CCAGAGAATGGTCTGGCACCCTTTC
Phenotype
MIM: 606994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929697 PubMed Links

Gene Details

Gene
LOC101929697
Gene Name
uncharacterized LOC101929697
There are no transcripts associated with this gene.

Gene
TNK2
Gene Name
tyrosine kinase non receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010938.1 4280 UTR 3 NP_001010938.1
NM_001308046.1 4280 UTR 3 NP_001294975.1
NM_005781.4 4280 UTR 3 NP_005772.3
XM_005269268.3 4280 UTR 3 XP_005269325.1
XM_005269270.3 4280 UTR 3 XP_005269327.1
XM_005269274.3 4280 UTR 3 XP_005269331.1
XM_005269275.3 4280 UTR 3 XP_005269332.1
XM_011512317.2 4280 UTR 3 XP_011510619.1
XM_011512318.2 4280 UTR 3 XP_011510620.2
XM_011512320.1 4280 UTR 3 XP_011510622.1
XM_011512321.2 4280 UTR 3 XP_011510623.1
XM_017005508.1 4280 UTR 3 XP_016860997.1
XM_017005509.1 4280 Intron XP_016860998.1
XM_017005510.1 4280 UTR 3 XP_016860999.1

View Full Product Details