Product Details

SNP ID
rs139778759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172444763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAGCTAGTTTTTAAAAGTTAC[A/G]TTTTAAATTTTGTATTGTAATGTCA
Phenotype
MIM: 601898
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GHSR PubMed Links

Gene Details

Gene
GHSR
Gene Name
growth hormone secretagogue receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004122.2 1542 Intron NP_004113.1
NM_198407.2 1542 UTR 3 NP_940799.1

View Full Product Details