Product Details

SNP ID
rs140128010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39098434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTGGTCCGCCTGGGCTTGGGC[A/C]CCTGGGCTGTCCAGGAAGGAGACCT
Phenotype
MIM: 606867 MIM: 611430 MIM: 612167
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GORASP1 PubMed Links

Gene Details

Gene
GORASP1
Gene Name
golgi reassembly stacking protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278789.1 1867 Silent Mutation GGG,GGT G280G NP_001265718.1
NM_001278790.1 1867 Silent Mutation GGG,GGT G220G NP_001265719.1
NM_031899.3 1867 Silent Mutation GGG,GGT G375G NP_114105.1
XM_006713301.3 1867 Silent Mutation GGG,GGT G380G XP_006713364.1
XM_011534020.2 1867 Silent Mutation GGG,GGT G350G XP_011532322.1
XM_011534021.2 1867 Silent Mutation GGG,GGT G320G XP_011532323.1
XM_017007050.1 1867 Silent Mutation GGG,GGT G345G XP_016862539.1
XM_017007051.1 1867 Silent Mutation GGG,GGT G315G XP_016862540.1
XM_017007052.1 1867 Silent Mutation GGG,GGT G218G XP_016862541.1
Gene
TTC21A
Gene Name
tetratricopeptide repeat domain 21A
There are no transcripts associated with this gene.

Gene
WDR48
Gene Name
WD repeat domain 48
There are no transcripts associated with this gene.

View Full Product Details