Product Details

SNP ID
rs141338964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49887330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAAGTGGGCCGAGGAGGCTCTG[A/T]GAGTGGCCGGGGCCGGCGTACATTC
Phenotype
MIM: 600168
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MST1R PubMed Links

Gene Details

Gene
MST1R
Gene Name
macrophage stimulating 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244937.2 4206 Missense Mutation ACA,TCA T1345S NP_001231866.1
NM_001318913.1 4206 Missense Mutation ACA,TCA T1288S NP_001305842.1
NM_002447.3 4206 Missense Mutation ACA,TCA T1394S NP_002438.2
XM_005265170.4 4206 Missense Mutation ACA,TCA T1395S XP_005265227.2
XM_011533739.2 4206 Missense Mutation ACA,TCA T1355S XP_011532041.1
XM_011533740.2 4206 Missense Mutation ACA,TCA T1346S XP_011532042.1
XM_011533741.2 4206 Missense Mutation ACA,TCA T1326S XP_011532043.1
XM_011533742.2 4206 UTR 3 XP_011532044.1
XM_011533743.2 4206 Missense Mutation ACA,TCA T1294S XP_011532045.1
XM_011533744.2 4206 Missense Mutation ACA,TCA T1289S XP_011532046.1

View Full Product Details