Product Details

SNP ID
rs141354129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27375166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGAATTTAAAACACCAGACTTG[C/T]TAAAATTTCAGCTTTTGGCATTCCT
Phenotype
MIM: 603353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEK10 PubMed Links

Gene Details

Gene
NEK10
Gene Name
NIMA related kinase 10
There are no transcripts associated with this gene.

Gene
SLC4A7
Gene Name
solute carrier family 4 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258379.1 4942 UTR 3 NP_001245308.1
NM_001258380.1 4942 UTR 3 NP_001245309.1
NM_001321103.1 4942 UTR 3 NP_001308032.1
NM_001321104.1 4942 UTR 3 NP_001308033.1
NM_001321105.1 4942 UTR 3 NP_001308034.1
NM_001321106.1 4942 UTR 3 NP_001308035.1
NM_001321107.1 4942 UTR 3 NP_001308036.1
NM_001321108.1 4942 UTR 3 NP_001308037.1
NM_003615.4 4942 UTR 3 NP_003606.3
XM_005265598.4 4942 UTR 3 XP_005265655.1
XM_005265600.4 4942 Intron XP_005265657.1
XM_006713421.2 4942 Intron XP_006713484.1
XM_011534256.2 4942 Intron XP_011532558.1
XM_011534258.1 4942 UTR 3 XP_011532560.1
XM_011534261.2 4942 UTR 3 XP_011532563.1
XM_011534263.2 4942 Intron XP_011532565.1
XM_011534265.1 4942 UTR 3 XP_011532567.1
XM_017007526.1 4942 Intron XP_016863015.1
XM_017007527.1 4942 Intron XP_016863016.1
XM_017007528.1 4942 UTR 3 XP_016863017.1
XM_017007529.1 4942 UTR 3 XP_016863018.1

View Full Product Details