Product Details

SNP ID
rs142424675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131535199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAAAACTGTGTGGGGAGTTCAT[A/G]GTGCTAGTGTTCTGGAAGATTCATA
Phenotype
MIM: 604208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L12P1 PubMed Links

Gene Details

Gene
BCL2L12P1
Gene Name
BCL2 like 12 pseudogene 1
There are no transcripts associated with this gene.

Gene
CPNE4
Gene Name
copine 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289112.1 3300 Missense Mutation CCA,CTA P575L NP_001276041.1
NM_130808.2 3300 Missense Mutation CCA,CTA P557L NP_570720.1
NM_153429.1 3300 Missense Mutation CCA,CTA P575L NP_702907.1
XM_011512406.2 3300 Missense Mutation CCA,CTA P444L XP_011510708.1
XM_011512407.2 3300 Missense Mutation CCA,CTA P444L XP_011510709.1
XM_011512408.2 3300 Missense Mutation CCA,CTA P444L XP_011510710.1
XM_017005693.1 3300 Missense Mutation CCA,CTA P575L XP_016861182.1
XM_017005694.1 3300 Intron XP_016861183.1
XM_017005695.1 3300 Missense Mutation CCA,CTA P417L XP_016861184.1
XM_017005696.1 3300 Missense Mutation CCA,CTA P306L XP_016861185.1

View Full Product Details