Product Details

SNP ID
rs143063772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42908888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCGGGGAGCTCTGGATGGAGAG[A/G]CCCCAAGGGGCATCTCCTCAGGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF662 PubMed Links

Gene Details

Gene
ZNF662
Gene Name
zinc finger protein 662
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134656.1 496 Missense Mutation ACC,GCC T104A NP_001128128.1
NM_207404.3 496 Missense Mutation ACC,GCC T44A NP_997287.2

View Full Product Details