Product Details

SNP ID
rs143243565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52247725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGGAGGCCTCAGGCCAGATGG[A/G]CGGCTGCACAGCCCTGGTGGCTGTG
Phenotype
MIM: 608979 MIM: 607433 MIM: 611059
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929054 PubMed Links

Gene Details

Gene
LOC101929054
Gene Name
uncharacterized LOC101929054
There are no transcripts associated with this gene.

Gene
PPM1M
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122870.2 562 Missense Mutation GAC,GGC D2G NP_001116342.1
NM_144641.3 562 Missense Mutation GAC,GGC D214G NP_653242.3
XM_005264879.2 562 Missense Mutation GAC,GGC D214G XP_005264936.1
XM_005264880.3 562 Missense Mutation GAC,GGC D129G XP_005264937.1
XM_005264881.1 562 Missense Mutation GAC,GGC D53G XP_005264938.1
Gene
TWF2
Gene Name
twinfilin actin binding protein 2
There are no transcripts associated with this gene.

Gene
WDR82
Gene Name
WD repeat domain 82
There are no transcripts associated with this gene.

View Full Product Details