Product Details

SNP ID
rs143852427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131382140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACGGGCTGAACCGCGAGCTGC[A/G]CGAGGAGCTGGGCGAAGCGGCTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC339874 PubMed Links

Gene Details

Gene
LOC339874
Gene Name
uncharacterized LOC339874
There are no transcripts associated with this gene.

Gene
NUDT16
Gene Name
nudix hydrolase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171905.1 170 Missense Mutation CAC,CGC H32R NP_001165376.1
NM_001171906.1 170 Missense Mutation CAC,CGC H78R NP_001165377.1
NM_152395.2 170 Missense Mutation CAC,CGC H78R NP_689608.2

View Full Product Details