Product Details

SNP ID
rs144472155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48620918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATGACGTAGGTGGCCTTCGATG[C/T]GGACCAGGAGGGCATCCAGCATGTG
Phenotype
MIM: 610068
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC26A6 PubMed Links

Gene Details

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
There are no transcripts associated with this gene.

Gene
TMEM89
Gene Name
transmembrane protein 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008269.1 404 Missense Mutation CAC,CGC H135R NP_001008270.1

View Full Product Details