Product Details
- SNP ID
-
rs145326060
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:49163175 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCTTGGCCCGTACTGCCTTGGCT[C/T]TAGCCTTGGCCTTGGCCCATGCTGC
- Phenotype
-
MIM: 613169
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC71
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4955418] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC71
- Gene Name
- coiled-coil domain containing 71
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022903.3 |
1172 |
Missense Mutation |
AAA,AGA |
K345R |
NP_075054.3 |
- Gene
- KLHDC8B
- Gene Name
- kelch domain containing 8B
There are no transcripts associated with this gene.
- Gene
- LAMB2P1
- Gene Name
- laminin subunit beta 2 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details