Product Details

SNP ID
rs145960363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125447811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTAAAGCATTCCTTAGCATTGG[C/T]CCAAACTTGAATTCCCTAAAAATAA
Phenotype
MIM: 605931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX4 PubMed Links

Gene Details

Gene
SNX4
Gene Name
sorting nexin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003794.3 1451 Missense Mutation ACC,GCC T441A NP_003785.1
XM_017007414.1 1451 Missense Mutation ACC,GCC T473A XP_016862903.1

View Full Product Details