Product Details

SNP ID
rs146138020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50331721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCAGGTAAAAGGAAGTGCGGC[A/G]CCTGACACTTGTGCCCCGTCCTGGG
Phenotype
MIM: 603551 MIM: 605082 MIM: 607052 MIM: 607070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYAL2 PubMed Links

Gene Details

Gene
HYAL2
Gene Name
hyaluronoglucosaminidase 2
There are no transcripts associated with this gene.

Gene
RASSF1
Gene Name
Ras association domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206957.1 637 Missense Mutation CGC,TGC R49C NP_001193886.1
NM_007182.4 637 Missense Mutation CGC,TGC R200C NP_009113.3
NM_170712.2 637 Missense Mutation CGC,TGC R49C NP_733830.1
NM_170713.2 637 Missense Mutation CGC,TGC R130C NP_733831.1
NM_170714.1 637 Missense Mutation CGC,TGC R204C NP_733832.1
XM_011533315.1 637 Missense Mutation CGC,TGC R130C XP_011531617.1
XM_011533316.2 637 Missense Mutation CGC,TGC R49C XP_011531618.1
Gene
RASSF1-AS1
Gene Name
RASSF1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TUSC2
Gene Name
tumor suppressor candidate 2
There are no transcripts associated with this gene.

Gene
ZMYND10
Gene Name
zinc finger MYND-type containing 10
There are no transcripts associated with this gene.

View Full Product Details