Product Details
- SNP ID
-
rs147390068
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:127639410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCTGGGCTGGGAGCCCCTGGCT[C/T]AGGCTTCCCCAGCGAAGAGAATGAA
- Phenotype
-
MIM: 616627
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PODXL2
PubMed Links
Gene Details
- Gene
- PODXL2
- Gene Name
- podocalyxin like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015720.3 |
297 |
Missense Mutation |
TCA,TTA |
S79L |
NP_056535.1 |
View Full Product Details