Product Details

SNP ID
rs147463121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48466679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGGCTCGCAGGCCCTGCCCCC[A/G]GGGCCCATGCAGACCCTCATCTTTT
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
There are no transcripts associated with this gene.

Gene
TREX1
Gene Name
three prime repair exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007248.3 1160 UTR 5 NP_009179.2
NM_016381.5 1160 Silent Mutation CCA,CCG P63P NP_057465.1
NM_033629.4 1160 Silent Mutation CCA,CCG P8P NP_338599.1

View Full Product Details