Product Details

SNP ID
hCV169749343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50286529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGGGCCATTGGCCCCCAACCA[C/T]GTGCATGAGGTATGCCTGCACCAGG
Phenotype
MIM: 604038 MIM: 602725 MIM: 607073 MIM: 601281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HYAL3 PubMed Links

Gene Details

Gene
HYAL3
Gene Name
hyaluronoglucosaminidase 3
There are no transcripts associated with this gene.

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
There are no transcripts associated with this gene.

Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
There are no transcripts associated with this gene.

Gene
NAT6
Gene Name
N-acetyltransferase 6
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
There are no transcripts associated with this gene.

View Full Product Details