Product Details
- SNP ID
-
rs148314795
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108757098 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATCCTTGATCTTCTTATCCATAA[C/T]GGAGTCTAAGGAACACTGAGTACTC
- Phenotype
-
MIM: 605645
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RETNLB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5851607] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RETNLB
- Gene Name
- resistin like beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032579.2 |
186 |
Missense Mutation |
ATT,GTT |
I30V |
NP_115968.1 |
View Full Product Details