Product Details

SNP ID
rs150437674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131382112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCGTGGACACGCAGGACAGAAGC[C/G]TAGAGGACGGGCTGAACCGCGAGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC339874 PubMed Links

Gene Details

Gene
LOC339874
Gene Name
uncharacterized LOC339874
There are no transcripts associated with this gene.

Gene
NUDT16
Gene Name
nudix hydrolase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171905.1 142 Missense Mutation CTA,GTA L23V NP_001165376.1
NM_001171906.1 142 Missense Mutation CTA,GTA L69V NP_001165377.1
NM_152395.2 142 Missense Mutation CTA,GTA L69V NP_689608.2

View Full Product Details