Product Details
- SNP ID
-
rs137965399
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:68053940 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGTCTTTGAGGCAATCCAATCT[C/T]GATACTTAGTTACTCTGGTGTAGAC
- Phenotype
-
MIM: 610892
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC550113
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10018943] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC550113
- Gene Name
- uncharacterized LOC550113
There are no transcripts associated with this gene.
- Gene
- SYT14P1
- Gene Name
- synaptotagmin 14 pseudogene 1
There are no transcripts associated with this gene.
- Gene
- TMPRSS11F
- Gene Name
- transmembrane protease, serine 11F
View Full Product Details