Product Details
- SNP ID
-
rs139222024
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:30721518 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCAGCCTGGCGGCCGCCAAGCA[A/G]CTCCTCCGGTACCGGCTGGCCGAGG
- Phenotype
-
MIM: 602988
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PCDH7
PubMed Links
Gene Details
- Gene
- PCDH7
- Gene Name
- protocadherin 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001173523.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
NP_001166994.1 |
NM_002589.2 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
NP_002580.2 |
NM_032456.2 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
NP_115832.1 |
NM_032457.3 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
NP_115833.2 |
XM_005248163.3 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_005248220.1 |
XM_005248164.3 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_005248221.1 |
XM_005248166.3 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_005248223.1 |
XM_011513842.2 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_011512144.1 |
XM_011513843.2 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_011512145.1 |
XM_011513844.2 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_011512146.1 |
XM_017008259.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863748.1 |
XM_017008260.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863749.1 |
XM_017008261.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863750.1 |
XM_017008262.1 |
1140 |
Missense Mutation |
CAA,CAG |
Q32Q |
XP_016863751.1 |
XM_017008263.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863752.1 |
XM_017008264.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863753.1 |
XM_017008265.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863754.1 |
XM_017008266.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863755.1 |
XM_017008267.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863756.1 |
XM_017008268.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863757.1 |
XM_017008269.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863758.1 |
XM_017008270.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863759.1 |
XM_017008271.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863760.1 |
XM_017008272.1 |
1140 |
Silent Mutation |
CAA,CAG |
Q32Q |
XP_016863761.1 |
View Full Product Details