Product Details
- SNP ID
-
rs139803589
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:55080163 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGCTTTTGCTGGGCACCATTCC[A/T]CTGCAGAAGAAATGGCAAACAAAGG
- Phenotype
-
MIM: 191306
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KDR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10006115] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KDR
- Gene Name
- kinase insert domain receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002253.2 |
4151 |
Silent Mutation |
GGA,GGT |
G1283G |
NP_002244.1 |
View Full Product Details